Assay Details
| Gene Symbol: Sf3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031179.2 | NP_112456.2 | 20 - 21 | 3123 | 84 | |
| GenBank mRNA | AB037890.1 | - | 20 - 21 | 3148 | 84 | |
| CF952155.1 | - | 2 - 3 | 188 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sf3b1
Genomic Map