Assay Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199293.1 | NP_001186222.1 | 22 - 23 | 3644 | 77 | |
| NM_031386.2 | NP_113563.2 | 22 - 23 | 3625 | 77 | ||
| GenBank mRNA | AK139808.1 | - | 9 - 10 | 1381 | 77 | |
More Information
Set Membership: |
|

Tex14
Genomic Map