Assay Details
| Gene Symbol: Nlrp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031389.2 | NP_113566.2 | 3 - 4 | 1976 | 69 | |
| GenBank mRNA | AF285581.1 | - | 1 - 2 | 1244 | 69 | |
| AK133284.1 | - | 3 - 4 | 1978 | 69 | ||
| AK166817.1 | - | 3 - 4 | 1957 | 69 | ||
| AK166869.1 | - | 3 - 4 | 2202 | 69 | ||
| AY596194.1 | - | 3 - 4 | 1976 | 69 | ||
| BC120833.1 | - | 1 - 2 | 1237 | 40129756 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105195 | AY596194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105195 | BC120833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105195 | AK166817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105195 | AK166869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105195 | AK133284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016882 | AY596194.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016882 | AK166817.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016882 | AK133284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016882 | NM_031389.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016882 | AF285581.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811880 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 54967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167095 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137209 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172234 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120524 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016882 | AY596194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016882 | AK166817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016882 | AK133284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016882 | NM_031389.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016882 | AF285581.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105195 | AY596194.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105195 | BC120833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105195 | AK166817.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105195 | AK166869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105195 | AK133284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234763 | AY596194.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234763 | BC120833.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234763 | AK166817.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234763 | AK133284.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234763 | NM_031389.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137209 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172234 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06116236 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120524 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06116248 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06116177 | - | - |
| Whole Mouse Genome G4122A | A_51_P234763 | AY596194.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234763 | BC120833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234763 | AK166817.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234763 | AK133284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P234763 | NM_031389.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137801 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132802 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129285 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172234 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137209 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24954033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24953862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06351624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06503174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06503173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06360346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05913167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06071850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06289559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06071846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06580889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06360362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24953820 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549805 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549808 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549811 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549807 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549806 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450000_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560746 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549804 | - | - |
| Mouse Genome 430 2.0 Array | 1450000_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450000_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83564_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83564_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000742.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000742.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000742.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000745.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000745.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000745.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000746.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000746.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000746.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000739.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000739.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000739.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000740.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000740.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000740.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000738.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000738.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000738.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000741.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000741.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000741.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000743.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000743.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000743.mm.1 | NM_031389.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000744.mm.1 | TC0700000066.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000744.mm.1 | NM_031389.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000744.mm.1 | NM_031389.2 | Refseq |
| Mouse Expression Set 430 | 1450000_PM_at | - | - |
| Mouse Expression Set 430 | 1450000_at | - | - |
| Murine Genome U74v2 Set | 131141_PM_at | - | - |
| Murine Genome U74v2 Set | 131141_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450000_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450000_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450000_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450000_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 466617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2680497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70524 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070524 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612942 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649947 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214165 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2612942 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214165 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649947 | - | - |
| mouse-6 v1.1 (Illumina) | scl0083564.2_63-S | - | - |
| mouse-6 v1.1 (Illumina) | scl083564.2_26-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0083564.2_153-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70524 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170184 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0083564.2_153-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0083564.2_63-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl083564.2_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170184 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680497 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70524 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2935933 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214165 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649947 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2612942 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612942 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2935933 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649947 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214165 | - | - |

Nlrp4c
Genomic Map