Assay Details
| Gene Symbol: Wdr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031392.2 | NP_113569.1 | 2 - 3 | 2660 | 81 | |
| GenBank mRNA | AB041854.1 | - | 2 - 3 | 2726 | 81 | |
| AF348591.1 | - | 2 - 3 | 2656 | 81 | ||
| AK037181.1 | - | 2 - 3 | 2637 | 81 | ||
| AK046119.1 | - | 2 - 3 | 2628 | 81 | ||
| AK052235.1 | - | 2 - 3 | 2601 | 81 | ||
| AK146617.1 | - | 2 - 3 | 2617 | 81 | ||
| AK163752.1 | - | 2 - 3 | 2593 | 81 | ||
| AK165935.1 | - | 2 - 3 | 2624 | 81 | ||
| AK166265.1 | - | 2 - 3 | 2638 | 81 | ||
| AK169357.1 | - | 2 - 3 | 2661 | 81 | ||
| AK205565.1 | - | 1 - 2 | 42 | 81 | ||
| BC035951.1 | - | 1 - 2 | 639 | 81 | ||
| BC050894.1 | - | 2 - 3 | 2660 | 5708880 | 81 | |
| BC054367.1 | - | 2 - 3 | 2588 | 5346943 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr6
Genomic Map