Assay Details
| Gene Symbol: Wdr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031392.2 | NP_113569.1 | 4 - 5 | 2873 | 58 | |
| GenBank mRNA | AB041854.1 | - | 4 - 5 | 2939 | 58 | |
| AF348591.1 | - | 4 - 5 | 2869 | 58 | ||
| AK037181.1 | - | 4 - 5 | 2850 | 58 | ||
| AK046119.1 | - | 4 - 5 | 2841 | 58 | ||
| AK052235.1 | - | 4 - 5 | 2814 | 58 | ||
| AK146617.1 | - | 4 - 5 | 2830 | 58 | ||
| AK163752.1 | - | 4 - 5 | 2806 | 58 | ||
| AK165935.1 | - | 4 - 5 | 2837 | 58 | ||
| AK166265.1 | - | 4 - 5 | 2851 | 58 | ||
| AK169357.1 | - | 4 - 5 | 2874 | 58 | ||
| AK177658.1 | - | 1 - 2 | 54 | 58 | ||
| AK187031.1 | - | 1 - 2 | 54 | 58 | ||
| AK215607.1 | - | 1 - 2 | 54 | 58 | ||
| BC035951.1 | - | 3 - 4 | 852 | 58 | ||
| BC050894.1 | - | 4 - 5 | 2873 | 5708880 | 58 | |
| BC054367.1 | - | 4 - 5 | 2801 | 5346943 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr6
Genomic Map