Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031393.2 | NP_113570.2 | 4 - 5 | 552 | 122 | |
| GenBank mRNA | AB050741.1 | - | 4 - 5 | 531 | 122 | |
| AK078868.1 | - | 4 - 5 | 541 | 123 | ||
| AK131162.1 | - | 4 - 5 | 509 | 122 | ||
| AK154176.1 | - | 4 - 5 | 547 | 122 | ||
| BC005623.1 | - | 4 - 5 | 552 | 3155535 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sytl1
Genomic Map