Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB050741.1 | - | 9 - 10 | 1037 | 87 | |
| AK154176.1 | - | 9 - 10 | 1054 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytl1| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB050741.1 | - | 9 - 10 | 1037 | 87 | |
| AK154176.1 | - | 9 - 10 | 1054 | 87 | ||
Set Membership: |
|