Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031393.2 | NP_113570.2 | 10 - 11 | 1161 | 71 | |
| GenBank mRNA | AB050741.1 | - | 10 - 11 | 1137 | 71 | |
| AK078868.1 | - | 10 - 11 | 1150 | 71 | ||
| AK131162.1 | - | 10 - 11 | 1118 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sytl1
Genomic Map