Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031393.2 | NP_113570.2 | 11 - 12 | 1321 | 72 | |
| GenBank mRNA | AB050741.1 | - | 11 - 12 | 1297 | 72 | |
| AK078868.1 | - | 11 - 12 | 1310 | 72 | ||
| AK131162.1 | - | 11 - 12 | 1278 | 72 | ||
| AK154176.1 | - | 11 - 12 | 1314 | 72 | ||
| BC005623.1 | - | 9 - 10 | 1873 | 3155535 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sytl1
Genomic Map