Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031393.2 | NP_113570.2 | 13 - 14 | 1504 | 71 | |
| GenBank mRNA | AB050741.1 | - | 13 - 14 | 1480 | 71 | |
| AK078868.1 | - | 13 - 14 | 1493 | 71 | ||
| AK131162.1 | - | 13 - 14 | 1461 | 71 | ||
| AK154176.1 | - | 13 - 14 | 1497 | 71 | ||
| BC005623.1 | - | 11 - 12 | 2056 | 3155535 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
synaptic vesicle exocytosis
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
protein binding
calcium-dependent phospholipid binding
Rab GTPase binding
syntaxin binding
clathrin binding
neurexin family protein binding
intracellular protein transport
exocytosis
synaptic vesicle exocytosis
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
protein binding
calcium-dependent phospholipid binding
Rab GTPase binding
syntaxin binding
clathrin binding
neurexin family protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163561 | AK078868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163561 | AK131162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163561 | NM_031393.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143693 | AK078868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143693 | AK131162.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143693 | NM_031393.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143693 | BC005623.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40462 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13248 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163561 | AK078868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163561 | AK131162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163561 | NM_031393.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143693 | AK078868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143693 | AK131162.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143693 | NM_031393.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143693 | BC005623.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485731 | AK154176.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485731 | BC005623.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485731 | AB050741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485731 | AK078868.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P485731 | AK131162.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23316673 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05408943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00992835 | - | - |
| Whole Mouse Genome G4122A | A_51_P485731 | AK154176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485731 | BC005623.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485731 | AB050741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485731 | AK078868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485731 | AK131162.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134502 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00992835 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03981736 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879775 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03461834 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03879782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23316673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03451412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03451411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04081315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03267568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03451405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04001440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23316628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04046863 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517011 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517010 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517022 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450240_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517009 | - | - |
| Mouse Genome 430 2.0 Array | 1450240_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450240_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269589_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269589_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400030263.mm.1 | TC0400003694.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030263.mm.1 | NM_031393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030263.mm.1 | NM_031393.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030266.mm.1 | TC0400003694.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030266.mm.1 | NM_031393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030266.mm.1 | NM_031393.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030264.mm.1 | TC0400003694.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030264.mm.1 | NM_031393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030264.mm.1 | NM_031393.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030265.mm.1 | TC0400003694.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030265.mm.1 | NM_031393.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030265.mm.1 | NM_031393.2 | Refseq |
| Mouse Expression Set 430 | 1450240_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450240_a_at | - | - |
| Murine Genome U74v2 Set | 129830_PM_g_at | - | - |
| Murine Genome U74v2 Set | 129829_PM_at | - | - |
| Murine Genome U74v2 Set | 129830_g_at | - | - |
| Murine Genome U74v2 Set | 129829_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450240_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450240_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450240_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450240_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 837857 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 510487 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 510487 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000510487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651054 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651054 | - | - |
| mouse-6 v1.1 (Illumina) | scl23718.13.1_59-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 510487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23718.13.1_59-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 510487 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2651054 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651054 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl23718.13.1_59-S | - | - |

Sytl1
Genomic Map