Assay Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040085.2 | NP_001035174.1 | 2 - 3 | 859 | 108 | |
| NM_001289583.1 | NP_001276512.1 | 2 - 3 | 859 | 108 | ||
| NM_001289584.1 | NP_001276513.1 | 2 - 3 | 859 | 108 | ||
| NM_031394.3 | NP_113571.2 | 2 - 3 | 859 | 108 | ||
| NR_110348.1 | 2 - 3 | 859 | 108 | |||
| GenBank mRNA | AB057754.1 | - | 1 - 2 | 198 | 108 | |
| AB057760.1 | - | 1 - 2 | 198 | 108 | ||
| AB057761.1 | - | 1 - 2 | 198 | 108 | ||
| AB057762.1 | - | 1 - 2 | 198 | 108 | ||
| BC052915.1 | - | 1 - 2 | 203 | 108 | ||
| BC064769.1 | - | 1 - 2 | 191 | 108 | ||
| BC138714.1 | - | 1 - 2 | 169 | 8861736 | 108 | |
| BC145398.1 | - | 1 - 2 | 169 | 9053956 | 108 | |
More Information
Set Membership: |
|

Sytl2
Genomic Map