Assay Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040085.2 | NP_001035174.1 | 6 - 7 | 1335 | 115 | |
| NM_001289583.1 | NP_001276512.1 | 6 - 7 | 1335 | 115 | ||
| NM_001289584.1 | NP_001276513.1 | 6 - 7 | 1254 | 115 | ||
| NM_031394.3 | NP_113571.2 | 6 - 7 | 1335 | 115 | ||
| NR_110348.1 | 6 - 7 | 1335 | 115 | |||
| GenBank mRNA | AB057754.1 | - | 5 - 6 | 674 | 115 | |
| AB057760.1 | - | 5 - 6 | 593 | 115 | ||
| AB057761.1 | - | 5 - 6 | 674 | 115 | ||
| AB057762.1 | - | 5 - 6 | 674 | 115 | ||
| BC052915.1 | - | 5 - 6 | 679 | 115 | ||
| BC064769.1 | - | 5 - 6 | 667 | 115 | ||
| BC138714.1 | - | 5 - 6 | 645 | 8861736 | 115 | |
| BC145398.1 | - | 5 - 6 | 645 | 9053956 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sytl2
Genomic Map