Assay Details
| Gene Symbol: Dbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031403.3 | NP_113580.2 | 5 - 6 | 841 | 71 | |
| GenBank mRNA | AK050090.1 | - | 5 - 6 | 842 | 71 | |
| AK090351.1 | - | 5 - 6 | 817 | 71 | ||
| AK139631.1 | - | 5 - 6 | 825 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dbr1
Genomic Map