Assay Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031406.3 | NP_113583.2 | 4 - 5 | 630 | 94 | |
| GenBank mRNA | AF314957.1 | - | 3 - 4 | 496 | 94 | |
| AK154118.1 | - | 4 - 5 | 588 | 94 | ||
| BC046982.1 | - | 4 - 5 | 550 | 6478046 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a9
Genomic Map