Assay Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031406.3 | NP_113583.2 | 7 - 8 | 1155 | 83 | |
| GenBank mRNA | AF314957.1 | - | 6 - 7 | 1021 | 83 | |
| AK154118.1 | - | 7 - 8 | 1114 | 83 | ||
| BC046982.1 | - | 7 - 8 | 1075 | 6478046 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a9
Genomic Map