Assay Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031406.3 | NP_113583.2 | 11 - 12 | 1718 | 68 | |
| GenBank mRNA | AF314957.1 | - | 10 - 11 | 1584 | 68 | |
| AK154118.1 | - | 11 - 12 | 1677 | 68 | ||
| BC046982.1 | - | 11 - 12 | 1638 | 6478046 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a9
Genomic Map