Assay Details
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031843.2 | NP_114031.2 | 4 - 5 | 526 | 55 | |
| GenBank mRNA | AF285235.1 | - | 4 - 5 | 521 | 55 | |
| AK140406.1 | - | 3 - 4 | 640 | 55 | ||
| AK140420.1 | - | 3 - 4 | 640 | 55 | ||
| AK154193.1 | - | 4 - 5 | 536 | 55 | ||
| BC027205.1 | - | 4 - 5 | 526 | 4190473 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp7
Genomic Map