Assay Details
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031843.2 | NP_114031.2 | 10 - 11 | 1240 | 81 | |
| GenBank mRNA | AF285235.1 | - | 10 - 11 | 1235 | 81 | |
| BC027205.1 | - | 10 - 11 | 1240 | 4190473 | 81 | |
| BC052861.1 | - | 7 - 8 | 1749 | 81 | ||
More Information
Set Membership: |
|

Dpp7
Genomic Map