Assay Details
| Gene Symbol: Dpp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031843.2 | NP_114031.2 | 11 - 12 | 1307 | 80 | |
| GenBank mRNA | AF285235.1 | - | 11 - 12 | 1302 | 80 | |
| AK028293.1 | - | 6 - 7 | 2667 | 80 | ||
| AK140406.1 | - | 8 - 9 | 2069 | 80 | ||
| AK140420.1 | - | 8 - 9 | 2070 | 80 | ||
| AK154193.1 | - | 10 - 11 | 1788 | 80 | ||
| BC027205.1 | - | 11 - 12 | 1307 | 4190473 | 80 | |
| BC052861.1 | - | 8 - 9 | 1816 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpp7
Genomic Map