Assay Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031863.3 | NP_114069.1 | 1 - 2 | 168 | 106 | |
| GenBank mRNA | AK011998.1 | - | 1 - 2 | 154 | 106 | |
| AK012179.1 | - | 1 - 2 | 169 | 106 | ||
| AK020040.1 | - | 1 - 2 | 170 | 106 | ||
| AK020761.1 | - | 1 - 2 | 664 | 106 | ||
| AK076094.1 | - | 1 - 2 | 146 | 106 | ||
| AK157569.1 | - | 1 - 2 | 196 | 106 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK011998.1,AK012179.1,AK020040.1,AK076094.1,NM_031863.3 transcripts.
Set Membership: |
|

Cenpq
Genomic Map