Assay Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031863.3 | NP_114069.1 | 2 - 3 | 302 | 93 | |
| GenBank mRNA | AK011998.1 | - | 2 - 3 | 288 | 93 | |
| AK012179.1 | - | 2 - 3 | 303 | 93 | ||
| AK020040.1 | - | 2 - 3 | 304 | 93 | ||
| AK020761.1 | - | 2 - 3 | 798 | 93 | ||
| AK076094.1 | - | 2 - 3 | 280 | 93 | ||
| AK088680.1 | - | 133 | 93 | |||
| AK157569.1 | - | 2 - 3 | 330 | 93 | ||
| BC018524.1 | - | 2 - 3 | 235 | 3588374 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpq
Genomic Map