Assay Details
| Gene Symbol: Cenpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031863.3 | NP_114069.1 | 3 - 4 | 362 | 129 | |
| GenBank mRNA | AK011998.1 | - | 3 - 4 | 348 | 129 | |
| AK012179.1 | - | 3 - 4 | 363 | 129 | ||
| AK020040.1 | - | 3 - 4 | 364 | 129 | ||
| AK020761.1 | - | 3 - 4 | 858 | 129 | ||
| AK076094.1 | - | 3 - 4 | 340 | 129 | ||
| AK088680.1 | - | 193 | 129 | |||
| BC018524.1 | - | 3 - 4 | 295 | 3588374 | 129 | |
More Information
Set Membership: |
|

Cenpq
Genomic Map