Assay Details
| Gene Symbol: Msh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282054.1 | NP_001268983.1 | 9 - 10 | 1360 | 107 | |
| NM_031870.3 | NP_114076.1 | 8 - 9 | 1451 | 107 | ||
| GenBank mRNA | AF178957.1 | - | 8 - 9 | 1155 | 107 | |
| AF298655.1 | - | 8 - 9 | 1451 | 107 | ||
| AK029837.1 | - | 9 - 10 | 1361 | 107 | ||
| AK161474.1 | - | 8 - 9 | 1201 | 107 | ||
| AY351585.1 | - | 8 - 9 | 1405 | 107 | ||
| BC138163.1 | - | 8 - 9 | 1342 | 8861182 | 107 | |
| BC145838.1 | - | 8 - 9 | 1342 | 40131141 | 107 | |
More Information
Set Membership: |
|

Msh4
Genomic Map