Assay Details
| Gene Symbol: Msh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282054.1 | NP_001268983.1 | 16 - 17 | 2237 | 75 | |
| NM_031870.3 | NP_114076.1 | 15 - 16 | 2328 | 75 | ||
| GenBank mRNA | AF178957.1 | - | 15 - 16 | 2032 | 75 | |
| AF298655.1 | - | 15 - 16 | 2328 | 75 | ||
| AK029837.1 | - | 16 - 17 | 2237 | 75 | ||
| AK161474.1 | - | 15 - 16 | 2078 | 75 | ||
| AY351585.1 | - | 15 - 16 | 2282 | 75 | ||
| AY351587.1 | - | 4 - 5 | 1279 | 75 | ||
| AY351590.1 | - | 7 - 8 | 1744 | 75 | ||
| AY351591.1 | - | 8 - 9 | 1768 | 75 | ||
| BC138163.1 | - | 15 - 16 | 2219 | 8861182 | 75 | |
| BC145838.1 | - | 15 - 16 | 2219 | 40131141 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Msh4
Genomic Map