| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NM_001159986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NM_001159986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NM_033354.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NM_033354.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NR_027641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
NR_027641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011723.mm.1 |
TC0100001434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NM_001159986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NM_001159986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NM_033354.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NM_033354.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NR_027641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
NR_027641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011722.mm.1 |
TC0100001434.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NM_001159986.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NM_001159986.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NM_033354.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NM_033354.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NR_027641.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
NR_027641.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011724.mm.1 |
TC0100001434.mm.1 |
MTA TC |