Assay Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033509.3 | NP_277044.1 | 2 - 3 | 536 | 95 | |
| GenBank mRNA | AF365875.1 | - | 2 - 3 | 300 | 95 | |
| AK033128.1 | - | 2 - 3 | 454 | 95 | ||
| AK046036.1 | - | 2 - 3 | 555 | 95 | ||
| AK049613.1 | - | 96 | ||||
| AK129311.1 | - | 536 | 95 | |||
| AY035370.3 | - | 2 - 3 | 524 | 95 | ||
| BC052195.1 | - | 2 - 3 | 536 | 6509008 | 95 | |
| BC075636.1 | - | 1393 | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Vangl2
Genomic Map