Assay Details
| Gene Symbol: Arid1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080819.1 | NP_001074288.1 | 12 - 13 | 3411 | 64 | |
| GenBank mRNA | AF268912.1 | - | 11 - 12 | 2262 | 64 | |
| BC082554.1 | - | 2643 | 30622048 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid1a
Genomic Map