Assay Details
| Gene Symbol: Amn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033603.2 | NP_291081.2 | 2 - 3 | 175 | 64 | |
| GenBank mRNA | AF320619.1 | - | 2 - 3 | 175 | 64 | |
| AK002318.1 | - | 2 - 3 | 174 | 64 | ||
| AK017204.1 | - | 2 - 3 | 189 | 64 | ||
| BC087954.1 | - | 2 - 3 | 188 | 30119116 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amn
Genomic Map