Assay Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053080.3 | NP_444310.3 | 5 - 6 | 604 | 85 | |
| GenBank mRNA | AF152359.1 | - | 5 - 6 | 559 | 85 | |
| AF246711.1 | - | 5 - 6 | 552 | 85 | ||
| AF253409.1 | - | 5 - 6 | 591 | 85 | ||
| AF280404.1 | - | 5 - 6 | 556 | 85 | ||
| AK043530.1 | - | 5 - 6 | 601 | 85 | ||
| AK086764.1 | - | 5 - 6 | 617 | 85 | ||
| AK147006.1 | - | 5 - 6 | 605 | 85 | ||
| BC026667.1 | - | 4 - 5 | 351 | 85 | ||
| BC058277.1 | - | 5 - 6 | 548 | 6515355 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1a3
Genomic Map