Assay Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053080.3 | NP_444310.3 | 7 - 8 | 850 | 67 | |
| GenBank mRNA | AF152359.1 | - | 7 - 8 | 805 | 67 | |
| AF246711.1 | - | 7 - 8 | 798 | 67 | ||
| AF253409.1 | - | 7 - 8 | 837 | 67 | ||
| AF280404.1 | - | 7 - 8 | 802 | 67 | ||
| AK147006.1 | - | 7 - 8 | 851 | 67 | ||
| BC058277.1 | - | 7 - 8 | 794 | 6515355 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh1a3
Genomic Map