Assay Details
| Gene Symbol: Fancg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163233.1 | NP_001156705.1 | 3 - 4 | 526 | 82 | |
| NM_053081.2 | NP_444311.1 | 3 - 4 | 526 | 82 | ||
| GenBank mRNA | AF112439.1 | - | 3 - 4 | 335 | 82 | |
| AY049715.1 | - | 3 - 4 | 326 | 82 | ||
| BC050890.1 | - | 4 - 5 | 630 | 82 | ||
| BC139763.1 | - | 3 - 4 | 329 | 40110334 | 82 | |
| BC141282.1 | - | 3 - 4 | 342 | 9055894 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fancg
Genomic Map