Assay Details
Gene Symbol: Pcdhb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053133.1 | NP_444363.1 | 1 | 2276 | 92 | |
GenBank mRNA | AY013790.1 | - | 1 | 2276 | 92 | |
BC116268.1 | - | 1 | 2277 | 92 | ||
BC116269.1 | - | 1 | 2269 | 92 | ||
BC127160.1 | - | 1 | 2276 | 40094058 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850529 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167968 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC116269.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC127160.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | AY013790.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC116268.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC116269.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC127160.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | AY013790.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | BC116268.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628029 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628009 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167968 | - | - |
Whole Mouse Genome G4122A | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | BC116269.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | BC127160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | AY013790.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | BC116268.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P302206 | NM_053133.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112259 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167968 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13928244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13928217 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13928254 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13928232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31627985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13994894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627995 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627983 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628021 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13746510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627987 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628007 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14131759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13928254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13885482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627977 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31627975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31628016 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10455079 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450341_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10455078 | - | - |
Mouse Genome 430 2.0 Array | 1450341_at | - | - |
Mouse Genome 430 2.0 Array | 1450341_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93879_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93879_PM_at | - | - |
Mouse Expression Set 430 | 1450341_PM_at | - | - |
Mouse Expression Set 430 | 1450341_at | - | - |
Mouse Genome 430A 2.0 Array | 1450341_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450341_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450341_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450341_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 406362 | - | - |
ABI Mouse Genome Survey Microarray | 411386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4010164 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4010164 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004010164 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242835 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242835 | - | - |
mouse-6 v1.1 (Illumina) | scl52066.1.1_263-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4010164 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52066.1.1_263-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4010164 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2648921 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2648921 | - | - |
