Assay Details
| Gene Symbol: Pcdhb19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053144.2 | NP_444374.2 | 1 | 305 | 83 | |
| GenBank mRNA | AF326312.1 | - | 1 | 149 | 83 | |
| AK029163.1 | - | 1 | 312 | 83 | ||
| AK046363.1 | - | 1 | 305 | 83 | ||
| AK147520.1 | - | 308 | 83 | |||
| AK160654.1 | - | 1 | 306 | 83 | ||
| AY013780.1 | - | 1 | 149 | 83 | ||
| BC141290.1 | - | 1 | 224 | 9055902 | 83 | |
| BC141291.1 | - | 1 | 224 | 9055903 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184730 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155471 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268469 | BC141291.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268469 | BC141290.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268469 | AK029163.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268469 | AK046363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268469 | AK147520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31628649 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155471 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08968331 | - | - |
| Whole Mouse Genome G4122A | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252953 | AK046363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252953 | AK147520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P252953 | NM_053144.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268469 | BC141291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268469 | BC141290.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268469 | AK029163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268469 | AK046363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268469 | AK147520.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139197 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915356 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915319 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915312 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13915338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31628679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14025026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13915333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13915320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628619 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31628643 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455124 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420443_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455123 | - | - |
| Mouse Genome 430 2.0 Array | 1420443_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420443_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93890_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 93890_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800002868.mm.1 | NM_053144.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002868.mm.1 | NM_053144.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002868.mm.1 | TC1800000376.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002867.mm.1 | NM_053144.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002867.mm.1 | NM_053144.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002867.mm.1 | TC1800000376.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420443_at | - | - |
| Mouse Expression Set 430 | 1420443_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420443_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420443_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420443_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420443_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 666145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105900576 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105900576 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2190411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747391 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2747391 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257036 | - | - |
| mouse-6 v1.1 (Illumina) | scl0093890.2_329-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52054.1_546-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105900576 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0093890.2_329-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190411 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105900576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2465874 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2465874 | - | - |

Pcdhb19
Genomic Map