| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NM_001040682.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NM_001040682.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NM_053155.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NM_053155.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NR_104435.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NR_104436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NR_104437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NR_104438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
NR_104439.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016028.mm.1 |
TC1200002296.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NM_001040682.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NM_001040682.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NM_053155.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NM_053155.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NR_104435.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NR_104436.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NR_104437.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NR_104438.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
NR_104439.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200016027.mm.1 |
TC1200002296.mm.1 |
MTA TC |