Assay Details
| Gene Symbol: Gopc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199272.1 | NP_001186201.1 | 6 - 7 | 1140 | 96 | |
| NM_053187.3 | NP_444417.2 | 5 - 6 | 1116 | 96 | ||
| GenBank mRNA | AB052838.1 | - | 5 - 6 | 896 | 96 | |
| AF287893.1 | - | 5 - 6 | 1032 | 96 | ||
| AK030427.1 | - | 6 - 7 | 736 | 96 | ||
| AK030637.1 | - | 6 - 7 | 949 | 96 | ||
| AK032613.1 | - | 6 - 7 | 946 | 96 | ||
| BC051171.1 | - | 5 - 6 | 1067 | 6307523 | 96 | |
More Information
Set Membership: |
|

Gopc
Genomic Map