Assay Details
| Gene Symbol: Cpsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164173.1 | NP_001157645.1 | 21 - 22 | 2489 | 62 | |
| NM_053193.2 | NP_444423.1 | 21 - 22 | 2489 | 62 | ||
| GenBank mRNA | AF322193.1 | - | 21 - 22 | 2460 | 62 | |
| AK149914.1 | - | 14 - 15 | 1488 | 62 | ||
| AK155806.1 | - | 21 - 22 | 2411 | 62 | ||
| BC011458.1 | - | 17 - 18 | 1925 | 62 | ||
| BC049941.1 | - | 21 - 22 | 2311 | 62 | ||
| BC056388.1 | - | 21 - 22 | 2489 | 6854851 | 62 | |
| BC057945.1 | - | 6 - 7 | 820 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf1
Genomic Map