Assay Details
| Gene Symbol: Foxp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286607.1 | NP_001273536.1 | 15 - 16 | 2091 | 75 | |
| NM_053242.4 | NP_444472.2 | 17 - 18 | 2491 | 75 | ||
| NM_212435.1 | NP_997600.1 | 15 - 16 | 2154 | 75 | ||
| GenBank mRNA | AF339106.1 | - | 14 - 15 | 1832 | 75 | |
| AK082361.1 | - | 17 - 18 | 2163 | 75 | ||
| AY079003.1 | - | 17 - 18 | 2120 | 75 | ||
| BC058960.1 | - | 15 - 16 | 2088 | 6811877 | 75 | |
| BC062926.1 | - | 15 - 16 | 2154 | 6851153 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxp2
Genomic Map