Assay Details
| Gene Symbol: Slc5a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053248.2 | NP_444478.2 | 1 - 2 | 447 | 55 | |
| GenBank mRNA | AF235001.1 | - | 1 - 2 | 414 | 55 | |
| AF380353.1 | - | 1 - 2 | 369 | 55 | ||
| AK046718.1 | - | 1 - 2 | 448 | 55 | ||
| BB645718.1 | - | 1 - 2 | 447 | 55 | ||
| BC137650.1 | - | 1 - 2 | 382 | 8860669 | 55 | |
| BC137651.1 | - | 1 - 2 | 382 | 8860670 | 55 | |
More Information
Set Membership: |
|

Slc5a5
Genomic Map