Assay Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053261.2 | NP_444491.1 | 2 - 3 | 419 | 84 | |
| GenBank mRNA | AF353730.1 | - | 2 - 3 | 418 | 84 | |
| AK154489.1 | - | 2 - 3 | 457 | 84 | ||
| AK154845.1 | - | 2 - 3 | 390 | 84 | ||
| AK170962.1 | - | 2 - 3 | 457 | 84 | ||
| AK171626.1 | - | 2 - 3 | 392 | 84 | ||
| BC011093.1 | - | 2 - 3 | 394 | 4012163 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impa2
Genomic Map