Assay Details
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054057.4 | NP_473398.1 | 6 - 7 | 658 | 85 | |
GenBank mRNA | AK032308.1 | - | 6 - 7 | 608 | 85 | |
AK134549.1 | - | 6 - 7 | 591 | 85 | ||
AK160172.1 | - | 6 - 7 | 612 | 85 | ||
AK211128.1 | - | 2 - 3 | 281 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
