Assay Details
| Gene Symbol: Mta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171052.1 | NP_001164523.1 | 5 - 6 | 475 | 74 | |
| NM_001171053.1 | NP_001164524.1 | 5 - 6 | 475 | 74 | ||
| NM_001171054.1 | NP_001164525.1 | 5 - 6 | 475 | 74 | ||
| NM_054082.2 | NP_473423.1 | 5 - 6 | 475 | 74 | ||
| GenBank mRNA | AF288138.1 | - | 5 - 6 | 429 | 74 | |
| AK129318.1 | - | 5 - 6 | 404 | 74 | ||
| AK145942.1 | - | 5 - 6 | 420 | 74 | ||
| AK146721.1 | - | 5 - 6 | 428 | 74 | ||
| AK146905.1 | - | 5 - 6 | 428 | 74 | ||
| AK147197.1 | - | 5 - 6 | 418 | 74 | ||
| AK154856.1 | - | 5 - 6 | 415 | 74 | ||
| AK158866.1 | - | 4 - 5 | 338 | 74 | ||
| BC022124.1 | - | 5 - 6 | 439 | 5097283 | 74 | |
| BC099960.1 | - | 5 - 6 | 392 | 30023887 | 74 | |
More Information
Set Membership: |
|

Mta3
Genomic Map