Assay Details
| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054085.2 | NP_473426.2 | 10 - 11 | 4366 | 84 | |
| GenBank mRNA | AF338872.1 | - | 10 - 11 | 4360 | 84 | |
| AK220384.1 | - | 6 - 7 | 2842 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpk3
Genomic Map