Assay Details
| Gene Symbol: Alpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054085.2 | NP_473426.2 | 12 - 13 | 4673 | 82 | |
| GenBank mRNA | AF338872.1 | - | 12 - 13 | 4667 | 82 | |
| AK052268.1 | - | 2 - 3 | 242 | 82 | ||
| AK220384.1 | - | 8 - 9 | 3149 | 82 | ||
More Information
Set Membership: |
|

Alpk3
Genomic Map