Assay Details
| Gene Symbol: Dpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058212.2 | NP_478119.1 | 3 - 4 | 332 | 79 | |
| GenBank mRNA | AF362750.1 | - | 3 - 4 | 304 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpf3
Genomic Map