Assay Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 3 - 4 | 219 | 74 | |
| NM_207706.1 | NP_997589.1 | 3 - 4 | 219 | 74 | ||
| GenBank mRNA | AF398884.1 | - | 2 - 3 | 124 | 74 | |
| AK032033.1 | - | 3 - 4 | 192 | 74 | ||
| AK033918.1 | - | 3 - 4 | 413 | 74 | ||
| AK038097.1 | - | 3 - 4 | 210 | 74 | ||
| AK038455.1 | - | 3 - 4 | 325 | 74 | ||
| AK045428.1 | - | 3 - 4 | 220 | 74 | ||
| AK047040.1 | - | 3 - 4 | 181 | 74 | ||
| AK050823.1 | - | 3 - 4 | 182 | 74 | ||
| AK053574.1 | - | 3 - 4 | 183 | 74 | ||
| AK129023.1 | - | 3 - 4 | 219 | 74 | ||
| BC023954.1 | - | 3 - 4 | 204 | 5320429 | 74 | |
| BC033418.1 | - | 2 - 3 | 149 | 74 | ||
More Information
Set Membership: |
|

Elmo2
Genomic Map