Assay Details
| Gene Symbol: Adcy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080435.1 | NP_536683.1 | 1 - 2 | 85 | 74 | |
| GenBank mRNA | AF442771.1 | - | 1 - 2 | 85 | 74 | |
| AK161880.1 | - | 1 - 2 | 104 | 74 | ||
| BC015299.1 | - | 1 - 2 | 85 | 4236007 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF442771.1,AK161880.1,BC015299.1,NM_080435.1 transcripts.
Set Membership: |
|

Adcy4
Genomic Map