Assay Details
| Gene Symbol: Adcy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080435.1 | NP_536683.1 | 21 - 22 | 2708 | 72 | |
| GenBank mRNA | AF442771.1 | - | 21 - 22 | 2708 | 72 | |
| AK141767.1 | - | 21 - 22 | 2779 | 72 | ||
| AK161880.1 | - | 21 - 22 | 2727 | 72 | ||
| AK220443.1 | - | 17 - 18 | 3470 | 72 | ||
| BC015299.1 | - | 21 - 22 | 2708 | 4236007 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adcy4
Genomic Map