Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 14 - 15 | 3248 | 88 | |
| GenBank mRNA | AB093303.1 | - | 14 - 15 | 3123 | 88 | |
| AB120565.1 | - | 14 - 15 | 3133 | 88 | ||
| AK037213.1 | - | 12 - 13 | 2958 | 88 | ||
| AK169543.1 | - | 15 - 16 | 3426 | 88 | ||
| BC008560.1 | - | 7 - 8 | 2221 | 88 | ||
| BC018526.1 | - | 4 - 5 | 452 | 88 | ||
| BC079535.1 | - | 8 - 9 | 2627 | 88 | ||
| BC122519.1 | - | 14 - 15 | 3134 | 88 | ||
| BC141883.1 | - | 14 - 15 | 3133 | 40097831 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssfa2
Genomic Map