Assay Details
| Gene Symbol: Rnf144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081977.2 | NP_001075446.1 | 2 - 3 | 234 | 62 | |
| GenBank mRNA | AF360998.1 | - | 2 - 3 | 230 | 62 | |
| BU055293.1 | - | 3 - 4 | 171 | 62 | ||
| BY323271.1 | - | 2 - 3 | 208 | 62 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF360998.1,NM_001081977.2 transcripts.
Set Membership: |
|

Rnf144a
Genomic Map