Assay Details
| Gene Symbol: Hars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077621.1 | - | 1 - 2 | 246 | 82 | |
| AK088789.1 | - | 1 - 2 | 264 | 82 | ||
| AK145003.1 | - | 1 - 2 | 251 | 82 | ||
| BC004596.1 | - | 1 - 2 | 210 | 3584555 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625831 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186562 | BC004596.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186562 | BC004665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186562 | AK145003.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186562 | AK077621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186562 | AK088789.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150662 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144408 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186562 | BC004596.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186562 | BC004665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186562 | AK145003.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186562 | AK077621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186562 | AK088789.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316431 | BC004596.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316431 | AK145003.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P316431 | NM_080636.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625877 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08967140 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08967155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144408 | - | - |
| Whole Mouse Genome G4122A | A_51_P316431 | BC004596.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316431 | AK145003.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316431 | NM_080636.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102475 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144408 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13602424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13602429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14119115 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625831 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13982559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14002039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13705893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14002040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13611978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13865528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13747260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13747262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13982564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14002035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10455003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455001 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419158_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449371_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454993 | - | - |
| Mouse Genome 430 2.0 Array | 1419158_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419158_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449371_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449371_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70791_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70791_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800002663.mm.1 | NM_080636.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002663.mm.1 | TC1800000346.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800002662.mm.1 | NM_080636.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800002662.mm.1 | TC1800000346.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419158_a_at | - | - |
| Mouse Expression Set 430 | 1449371_at | - | - |
| Mouse Expression Set 430 | 1419158_PM_a_at | - | - |
| Mouse Expression Set 430 | 1449371_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449371_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419158_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419158_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449371_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419158_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419158_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449371_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449371_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 553028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400215 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104150736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400215 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400215 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2675883 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2675883 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256363 | - | - |
| mouse-6 v1.1 (Illumina) | scl52079.13_121-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400215 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104150736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52079.13_121-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400215 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104150736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2675883 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256363 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2675883 | - | - |

Hars2
Genomic Map